siRNA Details
| Gene Symbol: RGD1561648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127567.1 | NP_001121039.1 | 2 | 321 | ||
| GenBank mRNA | BC166520.1 | - | 2 | 321 | 7133986 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1561648| Gene Symbol: RGD1561648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127567.1 | NP_001121039.1 | 2 | 321 | ||
| GenBank mRNA | BC166520.1 | - | 2 | 321 | 7133986 | |