siRNA Details
| Gene Symbol: RGD1565498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109287.2 | NP_001102757.2 | 3 | 953 | ||
| NM_001271273.1 | NP_001258202.1 | 3 | 786 | |||
| GenBank mRNA | FQ212465.1 | - | 3 | 786 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1565498| Gene Symbol: RGD1565498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109287.2 | NP_001102757.2 | 3 | 953 | ||
| NM_001271273.1 | NP_001258202.1 | 3 | 786 | |||
| GenBank mRNA | FQ212465.1 | - | 3 | 786 | ||