siRNA Details
| Gene Symbol: Dtx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191989.1 | NP_001178918.1 | 5, 6 | 1286 | ||
| XM_006241495.2 | XP_006241557.1 | 5, 6 | 2321 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dtx3| Gene Symbol: Dtx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191989.1 | NP_001178918.1 | 5, 6 | 1286 | ||
| XM_006241495.2 | XP_006241557.1 | 5, 6 | 2321 | |||