siRNA Details
| Gene Symbol: Dtx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191989.1 | NP_001178918.1 | 6 | 1342 | ||
| XM_006241492.2 | XP_006241554.1 | 5 | 2692 | |||
| XM_006241493.2 | XP_006241555.1 | 5 | 2534 | |||
| XM_006241494.2 | XP_006241556.1 | 5 | 2710 | |||
| XM_006241495.2 | XP_006241557.1 | 6 | 2377 | |||

Dtx3
Genomic Map