siRNA Details
| Gene Symbol: Zfp706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126087.1 | NP_001119559.1 | 2 | 210 | ||
| GenBank mRNA | BC083795.1 | - | 2 | 210 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp706| Gene Symbol: Zfp706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126087.1 | NP_001119559.1 | 2 | 210 | ||
| GenBank mRNA | BC083795.1 | - | 2 | 210 | ||