siRNA Details
| Gene Symbol: Rspo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130575.1 | NP_001124047.1 | 1, 2 | 1090 | ||
| XM_008765455.1 | XP_008763677.1 | 1, 2 | 539 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rspo2| Gene Symbol: Rspo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130575.1 | NP_001124047.1 | 1, 2 | 1090 | ||
| XM_008765455.1 | XP_008763677.1 | 1, 2 | 539 | |||