siRNA Details
| Gene Symbol: Sybu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241616.1 | XP_006241678.1 | 7, 8 | 1050 | ||
| XM_006241617.1 | XP_006241679.1 | 7, 8 | 1047 | |||
| XM_006241618.2 | XP_006241680.1 | 6, 7 | 1210 | |||
| XM_006241619.2 | XP_006241681.1 | 4, 5 | 1977 | |||
| XM_008765476.1 | XP_008763698.1 | 6, 7 | 991 | |||
| XM_008765477.1 | XP_008763699.1 | 6, 7 | 991 | |||
| XM_008765478.1 | XP_008763700.1 | 3, 4 | 366 | |||
| GenBank mRNA | FQ211706.1 | - | 7, 8 | 1047 | ||

Sybu
Genomic Map