siRNA Details
| Gene Symbol: Fam83a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130576.1 | NP_001124048.1 | 1 | 341 | ||
| XM_008765518.1 | XP_008763740.1 | 2 | 2976 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam83a| Gene Symbol: Fam83a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130576.1 | NP_001124048.1 | 1 | 341 | ||
| XM_008765518.1 | XP_008763740.1 | 2 | 2976 | |||