siRNA Details
| Gene Symbol: LOC500877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047963.1 | NP_001041428.1 | 4 | 312 | ||
| XM_006241694.1 | XP_006241756.1 | 3 | 402 | |||
| GenBank mRNA | AY325143.1 | - | 4 | 312 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC500877| Gene Symbol: LOC500877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047963.1 | NP_001041428.1 | 4 | 312 | ||
| XM_006241694.1 | XP_006241756.1 | 3 | 402 | |||
| GenBank mRNA | AY325143.1 | - | 4 | 312 | ||