siRNA Details
| Gene Symbol: Mfsd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024908.1 | NP_001020079.1 | 1 | 730 | ||
| XM_006241859.2 | XP_006241921.1 | 1 | 1231 | |||
| GenBank mRNA | BC097424.1 | - | 1 | 730 | 7455079 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mfsd3| Gene Symbol: Mfsd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024908.1 | NP_001020079.1 | 1 | 730 | ||
| XM_006241859.2 | XP_006241921.1 | 1 | 1231 | |||
| GenBank mRNA | BC097424.1 | - | 1 | 730 | 7455079 | |