siRNA Details
| Gene Symbol: Ncf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127304.1 | NP_001120776.1 | 6, 7 | 757 | ||
| XM_006241939.2 | XP_006242001.1 | 7, 8 | 1421 | |||
| GenBank mRNA | BC167076.1 | - | 6, 7 | 729 | 7129758 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncf4| Gene Symbol: Ncf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127304.1 | NP_001120776.1 | 6, 7 | 757 | ||
| XM_006241939.2 | XP_006242001.1 | 7, 8 | 1421 | |||
| GenBank mRNA | BC167076.1 | - | 6, 7 | 729 | 7129758 | |