siRNA Details
| Gene Symbol: Cyth4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130577.1 | NP_001124049.1 | 3 | 206 | ||
| XM_006242031.2 | XP_006242093.1 | 4 | 293 | |||
| XM_006242033.2 | XP_006242095.1 | 3 | 210 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyth4| Gene Symbol: Cyth4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130577.1 | NP_001124049.1 | 3 | 206 | ||
| XM_006242031.2 | XP_006242093.1 | 4 | 293 | |||
| XM_006242033.2 | XP_006242095.1 | 3 | 210 | |||