siRNA Details
| Gene Symbol: Msantd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024357.1 | NP_001019528.1 | 3 | 1197 | ||
| GenBank mRNA | BC096000.1 | - | 3 | 1197 | 7308051 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msantd4| Gene Symbol: Msantd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024357.1 | NP_001019528.1 | 3 | 1197 | ||
| GenBank mRNA | BC096000.1 | - | 3 | 1197 | 7308051 | |