siRNA Details
| Gene Symbol: RGD1561795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109289.1 | NP_001102759.1 | 2 | 295 | ||
| XM_008765967.1 | XP_008764189.1 | 4 | 634 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1561795| Gene Symbol: RGD1561795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109289.1 | NP_001102759.1 | 2 | 295 | ||
| XM_008765967.1 | XP_008764189.1 | 4 | 634 | |||