siRNA Details
| Gene Symbol: Zfp317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134634.2 | NP_001128106.2 | 7, 8 | 878 | ||
| XM_006242578.2 | XP_006242640.1 | 6, 7 | 870 | |||
| XM_008765976.1 | XP_008764198.1 | 4, 5 | 522 | |||
| XM_008765977.1 | XP_008764199.1 | 4, 5 | 498 | |||
| GenBank mRNA | BU671127.1 | - | 3, 4 | 248 | ||
| FQ211838.1 | - | 5, 6 | 665 | |||

Zfp317
Genomic Map