siRNA Details
Gene Symbol: Glb1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024358.1 | NP_001019529.2 | 8, 9 | 873 | ||
GenBank mRNA | BC083665.1 | - | 8, 9 | 873 | 7135489 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Glb1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024358.1 | NP_001019529.2 | 8, 9 | 873 | ||
GenBank mRNA | BC083665.1 | - | 8, 9 | 873 | 7135489 |