siRNA Details
| Gene Symbol: Slc37a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191994.1 | NP_001178923.1 | 4, 5 | 310 | ||
| XM_006242809.2 | XP_006242871.1 | 4, 5 | 409 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc37a2| Gene Symbol: Slc37a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191994.1 | NP_001178923.1 | 4, 5 | 310 | ||
| XM_006242809.2 | XP_006242871.1 | 4, 5 | 409 | |||