siRNA Details
| Gene Symbol: Tmem225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024359.1 | NP_001019530.1 | 2 | 381 | ||
| GenBank mRNA | AY634367.1 | - | 2 | 381 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem225| Gene Symbol: Tmem225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024359.1 | NP_001019530.1 | 2 | 381 | ||
| GenBank mRNA | AY634367.1 | - | 2 | 381 | ||