siRNA Details
| Gene Symbol: Cbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242965.2 | XP_006243027.1 | 4 | 766 | ||
| XM_576396.6 | XP_576396.3 | 4 | 766 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cbl| Gene Symbol: Cbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242965.2 | XP_006243027.1 | 4 | 766 | ||
| XM_576396.6 | XP_576396.3 | 4 | 766 | |||