siRNA Details
Gene Symbol: Ddx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109292.1 | NP_001102762.1 | 12, 13 | 1637 | ||
XM_006242950.2 | XP_006243012.1 | 12, 13 | 1637 | |||
XM_006242951.2 | XP_006243013.1 | 12, 13 | 1651 | |||
XM_006242952.2 | XP_006243014.1 | 12, 13 | 1646 | |||
XM_006242954.2 | XP_006243016.1 | 12, 13 | 1920 | |||
GenBank mRNA | FQ234306.1 | - | 4, 5 | 432 | ||
FQ235180.1 | - | 4, 5 | 422 |
