siRNA Details
| Gene Symbol: LOC500990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037364.1 | NP_001032441.1 | 7, 8 | 865 | ||
| XM_008766163.1 | XP_008764385.1 | 8, 9 | 1081 | |||
| XM_008766164.1 | XP_008764386.1 | 8, 9 | 1081 | |||
| XM_008766165.1 | XP_008764387.1 | 8, 9 | 1081 | |||
| XM_008766166.1 | XP_008764388.1 | 8, 9 | 1081 | |||
| XM_008766167.1 | XP_008764389.1 | 8, 9 | 1078 | |||
| XM_008766168.1 | XP_008764390.1 | 8, 9 | 1092 | |||
| XM_008766169.1 | XP_008764391.1 | 7, 8 | 998 | |||
| XM_008766170.1 | XP_008764392.1 | 7, 8 | 953 | |||
| GenBank mRNA | BC105843.1 | - | 7, 8 | 865 | 7455643 | |

LOC500990
Genomic Map