siRNA Details
| Gene Symbol: RGD1563941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109293.1 | NP_001102763.1 | 1 | 261 | ||
| GenBank mRNA | BC158857.1 | - | 372 | 7930388 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1563941| Gene Symbol: RGD1563941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109293.1 | NP_001102763.1 | 1 | 261 | ||
| GenBank mRNA | BC158857.1 | - | 372 | 7930388 | ||