siRNA Details
| Gene Symbol: RGD1562084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109294.1 | NP_001102764.1 | 1114 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562084| Gene Symbol: RGD1562084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109294.1 | NP_001102764.1 | 1114 | |||