siRNA Details
| Gene Symbol: RGD1562618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113791.1 | NP_001107263.1 | 2 | 1174 | ||
| GenBank mRNA | BC158644.1 | - | 2 | 1174 | 8363324 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562618| Gene Symbol: RGD1562618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113791.1 | NP_001107263.1 | 2 | 1174 | ||
| GenBank mRNA | BC158644.1 | - | 2 | 1174 | 8363324 | |