siRNA Details
Gene Symbol: Gfral | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191998.1 | NP_001178927.1 | 6 | 935 | ||
XM_006243416.2 | XP_006243478.1 | 7 | 2975 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gfral | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191998.1 | NP_001178927.1 | 6 | 935 | ||
XM_006243416.2 | XP_006243478.1 | 7 | 2975 |