siRNA Details
| Gene Symbol: LOC102554622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243468.2 | XP_006243530.1 | 2 | 336 | ||
| GenBank mRNA | FQ221412.1 | - | 1 | 32 | ||
| Gene Symbol: Sh3bgrl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137647.1 | NP_001131119.1 | 2 | 341 | ||

LOC102554622
Genomic Map