siRNA Details
| Gene Symbol: RGD1560775 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766460.1 | XP_008764682.1 | 16 | 1367 | ||
| XM_008766461.1 | XP_008764683.1 | 16 | 1106 | |||
| XM_008766462.1 | XP_008764684.1 | 16 | 1367 | |||
| XM_008766463.1 | XP_008764685.1 | 16 | 1367 | |||
| XM_008766464.1 | XP_008764686.1 | 15 | 1254 | |||
| XM_008766465.1 | XP_008764687.1 | 15 | 1175 | |||
| XM_008766466.1 | XP_008764688.1 | 16 | 1366 | |||
| GenBank mRNA | BC128758.1 | - | 15 | 949 | 7111588 | |

RGD1560775
Genomic Map