siRNA Details
| Gene Symbol: Plscr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191999.1 | NP_001178928.1 | 3 | 303 | ||
| XM_006243540.2 | XP_006243602.1 | 5 | 638 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plscr5| Gene Symbol: Plscr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191999.1 | NP_001178928.1 | 3 | 303 | ||
| XM_006243540.2 | XP_006243602.1 | 5 | 638 | |||