siRNA Details
Gene Symbol: Plscr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191999.1 | NP_001178928.1 | 4, 5 | 616 | ||
XM_006243540.2 | XP_006243602.1 | 6, 7 | 951 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Plscr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191999.1 | NP_001178928.1 | 4, 5 | 616 | ||
XM_006243540.2 | XP_006243602.1 | 6, 7 | 951 |