siRNA Details
| Gene Symbol: Il20rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173438.1 | NP_001166909.1 | 7 | 987 | ||
| GenBank mRNA | DQ222846.1 | - | 7 | 987 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il20rb| Gene Symbol: Il20rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173438.1 | NP_001166909.1 | 7 | 987 | ||
| GenBank mRNA | DQ222846.1 | - | 7 | 987 | ||