siRNA Details
| Gene Symbol: Poc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109296.1 | NP_001102766.1 | 6 | 744 | ||
| XM_008766561.1 | XP_008764783.1 | 7 | 1204 | |||
| XM_008766562.1 | XP_008764784.1 | 5 | 621 | |||
| GenBank mRNA | FQ233502.1 | - | 7 | 808 | ||

Poc1a
Genomic Map