siRNA Details
Gene Symbol: Gpr62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766596.1 | XP_008764818.1 | 1 | 3846 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
