siRNA Details
| Gene Symbol: Iqcf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025153.1 | NP_001020324.1 | 7, 8 | 646 | ||
| XM_006243837.1 | XP_006243899.1 | 7, 8 | 682 | |||
| GenBank mRNA | BC079282.1 | - | 7, 8 | 646 | 7134209 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iqcf3| Gene Symbol: Iqcf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025153.1 | NP_001020324.1 | 7, 8 | 646 | ||
| XM_006243837.1 | XP_006243899.1 | 7, 8 | 682 | |||
| GenBank mRNA | BC079282.1 | - | 7, 8 | 646 | 7134209 | |