siRNA Details
| Gene Symbol: Kif9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001192000.1 | NP_001178929.1 | 9 | 1027 | ||
| XM_006243994.2 | XP_006244056.1 | 10 | 1406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif9| Gene Symbol: Kif9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001192000.1 | NP_001178929.1 | 9 | 1027 | ||
| XM_006243994.2 | XP_006244056.1 | 10 | 1406 | |||