siRNA Details
| Gene Symbol: Prss44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109298.1 | NP_001102768.1 | 1 | 220 | ||
| XM_008766647.1 | XP_008764869.1 | 1 | 220 | |||
| GenBank mRNA | BC168977.1 | - | 1 | 199 | 7453880 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss44| Gene Symbol: Prss44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109298.1 | NP_001102768.1 | 1 | 220 | ||
| XM_008766647.1 | XP_008764869.1 | 1 | 220 | |||
| GenBank mRNA | BC168977.1 | - | 1 | 199 | 7453880 | |