siRNA Details
| Gene Symbol: Prss44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109298.1 | NP_001102768.1 | 3 | 664 | ||
| XM_008766647.1 | XP_008764869.1 | 3 | 664 | |||
| XM_008766648.1 | XP_008764870.1 | 3 | 340 | |||
| GenBank mRNA | BC168977.1 | - | 3 | 643 | 7453880 | |

Prss44
Genomic Map