siRNA Details
| Gene Symbol: Cmtm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109300.1 | NP_001102770.1 | 3 | 869 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cmtm7| Gene Symbol: Cmtm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109300.1 | NP_001102770.1 | 3 | 869 | ||