siRNA Details
| Gene Symbol: Pdcd6ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029910.3 | NP_001025081.2 | 13 | 1839 | ||
| XM_006243997.1 | XP_006244059.1 | 13 | 2174 | |||
| GenBank mRNA | AF192757.1 | - | 3 | 306 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdcd6ip| Gene Symbol: Pdcd6ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029910.3 | NP_001025081.2 | 13 | 1839 | ||
| XM_006243997.1 | XP_006244059.1 | 13 | 2174 | |||
| GenBank mRNA | AF192757.1 | - | 3 | 306 | ||