siRNA Details
| Gene Symbol: Tbc1d5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134762.1 | NP_001128234.1 | 5 | 483 | ||
| XM_008766768.1 | XP_008764990.1 | 5 | 1539 | |||
| XM_008766769.1 | XP_008764991.1 | 1 | 35 | |||
| GenBank mRNA | BC162056.1 | - | 5 | 483 | ||

Tbc1d5
Genomic Map