siRNA Details
| Gene Symbol: Slc35b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037215.3 | NP_001032292.1 | 4 | 564 | ||
| GenBank mRNA | AB197928.1 | - | 4 | 514 | ||
| BC100647.1 | - | 4 | 564 | 7932661 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc35b2| Gene Symbol: Slc35b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037215.3 | NP_001032292.1 | 4 | 564 | ||
| GenBank mRNA | AB197928.1 | - | 4 | 514 | ||
| BC100647.1 | - | 4 | 564 | 7932661 | ||