siRNA Details
Gene Symbol: Fam178b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122658.1 | NP_001116130.1 | 9 | 1273 | ||
XM_006244842.2 | XP_006244904.1 | 9 | 1294 | |||
XM_006244843.2 | XP_006244905.1 | 10 | 1408 | |||
XM_006244844.2 | XP_006244906.1 | 10 | 1365 | |||
XM_008767041.1 | XP_008765263.1 | 8 | 1040 | |||
GenBank mRNA | BC160833.1 | - | 9 | 1273 | 7456084 |
