siRNA Details
| Gene Symbol: Fam178b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122658.1 | NP_001116130.1 | 2 | 394 | ||
| XM_006244842.2 | XP_006244904.1 | 2 | 415 | |||
| XM_006244843.2 | XP_006244905.1 | 3 | 529 | |||
| XM_006244844.2 | XP_006244906.1 | 3 | 486 | |||
| XM_008767041.1 | XP_008765263.1 | 1 | 161 | |||
| GenBank mRNA | BC160833.1 | - | 2 | 394 | 7456084 | |

Fam178b
Genomic Map