siRNA Details
| Gene Symbol: Rfx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767059.1 | XP_008765281.1 | 13, 14 | 2621 | ||
| GenBank mRNA | FQ230624.1 | - | 8, 9 | 1182 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rfx8| Gene Symbol: Rfx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767059.1 | XP_008765281.1 | 13, 14 | 2621 | ||
| GenBank mRNA | FQ230624.1 | - | 8, 9 | 1182 | ||