siRNA Details
| Gene Symbol: Tmem182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109305.1 | NP_001102775.1 | 5 | 644 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem182| Gene Symbol: Tmem182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109305.1 | NP_001102775.1 | 5 | 644 | ||