siRNA Details
| Gene Symbol: Ankar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767142.1 | XP_008765364.1 | 509 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankar| Gene Symbol: Ankar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767142.1 | XP_008765364.1 | 509 | |||