siRNA Details
| Gene Symbol: LOC102550841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767304.1 | XP_008765526.1 | 5 | 550 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102550841| Gene Symbol: LOC102550841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767304.1 | XP_008765526.1 | 5 | 550 | ||