siRNA Details
| Gene Symbol: Vwc2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109308.1 | NP_001102778.1 | 4 | 740 | ||
| XM_006245123.2 | XP_006245185.1 | 2 | 1232 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vwc2l| Gene Symbol: Vwc2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109308.1 | NP_001102778.1 | 4 | 740 | ||
| XM_006245123.2 | XP_006245185.1 | 2 | 1232 | |||