siRNA Details
Gene Symbol: Vwc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109308.1 | NP_001102778.1 | 5 | 925 | ||
XM_006245123.2 | XP_006245185.1 | 3 | 1417 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vwc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109308.1 | NP_001102778.1 | 5 | 925 | ||
XM_006245123.2 | XP_006245185.1 | 3 | 1417 |