siRNA Details
| Gene Symbol: Mreg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001192002.1 | NP_001178931.1 | 2 | 213 | ||
| XM_006245265.2 | XP_006245327.1 | 3 | 361 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mreg| Gene Symbol: Mreg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001192002.1 | NP_001178931.1 | 2 | 213 | ||
| XM_006245265.2 | XP_006245327.1 | 3 | 361 | |||