siRNA Details
| Gene Symbol: Gpr55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006245491.2 | XP_006245553.1 | 2 | 2761 | ||
| XM_006245492.2 | XP_006245554.1 | 2 | 2379 | |||
| XM_006245493.2 | XP_006245555.1 | 2 | 1566 | |||
| XM_006245494.2 | XP_006245556.1 | 2 | 908 | |||
| XM_006245495.2 | XP_006245557.1 | 2 | 2497 | |||

Gpr55
Genomic Map